missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Matrix Gla Protein
A DNA sequence encoding the Homo sapiens (Human) Matrix Gla protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6359-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000891.2 | |
Matrix Gla Protein | |
Research Use Only | |
Mgp | |
Recombinant Protein | |
E. coli | |
YESHESMESY ELNPFINRRN ANTFISPQQR WRAKVQERIR ERSKPVHELN REACDDYRLC ERYAMVYGYN AAYNRYF | |
Greater than 90% as determined by SDS-PAGE. |
4256 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |