missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mannan-binding lectin serine protease 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Mannan-binding lectin serine protease 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6347-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002650

  • £1075.37 / 500µg

Estimated Shipment
09-11-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001027019.1
Mannan-binding lectin serine protease 1 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5648
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MASP1
Recombinant Protein
E. coli
HTVELNNMFG QIQSPGYPDS YPSDSEVTWN ITVPDGFRIK LYFMHFNLES SYLCEYDYVK VETEDQVLAT FCGRETTDTE QTPGQEVVLS PGSFMSITFR SDFSNEERFT GFDAHYMAVD VDECKEREDE ELSCDHYCHN YIGGYYCSCR FGYILHTDNR TCRVECSDNL FTQRTGVITS PDFPNPYPKS SECLYTIELE EGFMVNLQFE DIFDIEDHPE VPCPYDYIKI KVGPKVLGPF CGEKAPEPIS TQSHSVLILF HSDNSGENRG WRLSYRAAGN ECPELQPPVH GKIEPSQAKY FFKDQVLVSC DTGYKVLKDN VEMDTFQIEC LKDGTWSNKI PTCKIVDCRA PGELEHGLIT FSTRNNLTTY KSEIKYSCQE PYYKMLNNNT GIYTCSAQGV WMNKVLGRSL PTCLPVCGLP KFSRKLMARI FNGRPAQKGT TPWIAMLSHL NGQPFCGGSL LGSSWIVTAA HCLHQSLDPE DPTLRDSDLL SPSDFKIILG KHWRLRSDEN EQHLGVKHTT LHPQYDPNTF ENDVALVELL ESPVLNAFVM PICLPEGPQQ EGAMVIVSGW GKQFLQRFPE TLMEIEIPIV DHSTCQKAYA PLKKKVTRDM ICAGEKEGGK DACAGDSGGP MVTLNRERGQ WYLVGTVSWG DDCGKKDRYG VYSYIHHNKD WIQRVTGVRN
Videos
Documents