missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAFK Protein
A cDNA sequence encoding the MAFK was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12621-5ug
Additional Details : Weight : 0.01000kg
Specifications
MAFK Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTTNPKPNKA LKVKKEAGEN APVLSDDELV SMSVRELNQH LRGLTKEEVT RLKQRRRTLK NRGYAASCRI KRVTQKEELE RQRVELQQEV EKLARENSSM RLELDALRSK YEALQTFART VARGPVAPSK VATTSVITIV KSTELSSTSV PFSAAS | |
Greater than 85.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MAFK protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |