missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAFG Protein
A cDNA sequence encoding the MAFG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12620-2ug
Additional Details : Weight : 0.01000kg
Specifications
MAFG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTTPNKGNKA LKVKREPGEN GTSLTDEELV TMSVRELNQH LRGLSKEEIV QLKQRRRTLK NRGYAASCRV KRVTQKEELE KQKAELQQEV EKLASENASM KLELDALRSK YEALQTFART VARSPVAPAR GPLAAGLGPL VPGKVAATSV ITIVKSKTDA RS | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAFG solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.15M NaCl and 1mM DTT. |