missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LYVE1 Protein
A cDNA sequence encoding the LYVE1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12609-0.1mg
Additional Details : Weight : 0.01000kg
Specifications
10894 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DPLRAEELS IQVSCRIMGI TLVSKKANQQ LNFTEAKEAC RLLGLSLAGK DQVETALKAS FETCSYGWVG DGFVVISRIS PNPKCGKNGV GVLIRKVPVS RQFAAYCYNS SDTWTNSCIP EIITTKDPIF NTQTATQTTE FIVSDSTYSV ASPYSTIPAP TTTPPAPAST SIPRRKKLIC VTEVFMETST MSTETEPFVE NKAAFKNEAA GFGGSGRLVP RGSHHHHHH | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
LYVE1 Protein | |
Research Use Only | |
LYVE1 | |
Human | |
Untagged | |
The LYVE1 protein solution contains 20mM Tris buffer pH 7.5 and 10% Glycerol. |