missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lysozyme-like protein 1

A DNA sequence encoding the Homo sapiens (Human) Lysozyme-like protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7748-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050561

  • £1735.00 / 1mg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_115906.3
Lysozyme-like protein 1
Research Use Only
LYZL1
Recombinant Protein
E. coli
MQDAPLSCLS PTKWSSVSSA DSTEKSASGA GTRNLPFQFC LRQALRMKAA GILTLIGCLV TGAESKIYTR CKLAKIFSRA GLDNYWGFSL GNWICMAYYE SGYNTTAQTV LDDGSIDYGI FQINSFAWCR RGKLKENNHC HVACSALITD DLTDAIICAR KIVKETQGMN YWQGWKKHCE GRDLSEWKKG CEVS
Greater than 90% as determined by SDS-PAGE.
84569
1 mg
Not determined.
Full Length of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers