missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LY-96 / ESOP-1 Protein

A DNA sequence encoding the Homo sapiens (Human) LY-96 / ESOP-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6326-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082490

  • £1735.00 / 1mg
Estimated Shipment: 16-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001182726.1
LY-96 / ESOP-1 Protein
Research Use Only
LY96
Recombinant Protein
E. coli
EAQKQYWVCN SSDASISYTY CDKMQYPISI NVNPCIELKR SKGLLHIFYI PRRDLKQLYF NLYITVNTMN LPKRKEVICR GSDDDYSFCR ALKGETVNTT ISFSFKGIKF SKGKYKCVVE AISGSPEEML FCLEFVILHQ PNSN
Greater than 80% as determined by SDS-PAGE.
23643
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers