missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LTF Holo Protein

A cDNA sequence encoding the LTF Holo was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12596-1gram

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960939

  • £662.78 / 1g
Estimated Shipment: 04-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4057
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
LTF
Human
Untagged
The Human Holo lactotransferrin was lyophilized from water, contains 1.66 mg iron/gram Lactoferrin.
LTF Holo Protein
1 g
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Rice
MKLVFLVLLF LGALGLCLAG RRRSVQWCAV SQPEATKCFQ WQRNMRKVRG PPVSCIKRDS PIQCIQAIAE NRADAVTLDG GFIYEAGLAP YKLRPVAAEV YGTERQPRTH YYAVAVVKKG GSFQLNELQG LKSCHTGLRR TAGWNVPIGT LRPFLNWTGP PEPIEAAVAR FFSASCVPGA DKGQFPNLCR LCAGTGENKC AFSSQEPYFS YSGAFKCLRD GAGDVAFIRE STVFEDLSDE AERDEYELLC PDNTRKPVDK FKDCHLARVP SHAVVARSVN GKEDAIWNLL RQAQEKFGKD KSPKFQLFGS PSGQKDLLFK DSAIGFSRVP PRIDSGLYLG SGYFTAIQNL RKSEEEVAAR RARVVWCAVG EQELRKCNQW SGLSEGSVTC SSASTTEDCI ALVLKGEADA MSLDGGYVYT AGKCGLVPVL AENYKSQQSS DPDPNCVDRP VEGYLAVAVV RRSDTSLTWN SVKGKKSCHT AVDRTAGWNI PMGLLFNQTG SCKFDEYFSQ SCAPGSDPRS NLCALCIGDE QGENKCVPNS NERYYGYTGA FRCLAENAGD VAFVKDVTVL QNTDGNNNEA WAKDLKLADF ALLCLDGKRK PVTEARSCHL AMAPNHAVVS RMDKVERLKQ VLLHQQAKFG RNGSDCPDKF CLFQSETKNL LFNDNTECLA RLHGKTTYEK YLGPQYVAGI TNLKKCSTSP LLEACEFLRK
Videos
SDS
Documents
Certificates
Special Offers

Special Offers