missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LTA4H Protein
A cDNA sequence encoding the LTA4H was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12594-100ug
Additional Details : Weight : 0.01000kg
Specifications
4048 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPEIVDT CSLASPASVC RTKHLHLRCS VDFTRRTLTG TAALTVQSQE DNLRSLVLDT KDLTIEKVVI NGQEVKYALG ERQSYKGSPM EISLPIALSK NQEIVIEISF ETSPKSSALQ WLTPEQTSGK EHPYLFSQCQ AIHCRAILPC QDTPSVKLTY TAEVSVPKEL VALMSAIRDG ETPDPEDPSR KIYKFIQKVP IPCYLIALVV GALESRQIGP RTLVWSEKEQ VEKSAYEFSE TESMLKIAED LGGPYVWGQY DLLVLPPSFP YGGMENPCLT FVTPTLLAGD KSLSNVIAHE ISHSWTGNLV TNKTWDHFWL NEGHTVYLER HICGRLFGEK FRHFNALGGW GELQNSVKTF GETHPFTKLV VDLTDIDPDV AYSSVPYEKG FALLFYLEQL LGGPEIFLGF LKAYVEKFSY KSITTDDWKD FLYSYFKDKV DVLNQVDWNA WLYSPGLPPI KPNYDMTLTN ACIALSQRWI TAKEDDLNSF NATDLKDLSS HQLNEFLAQT LQRAPLPLGH IKRMQEVYNF NAINNSEIRF RWLRLCIQSK WEDAIPLALK MATEQGRMKF TRPLFKDLAA FDKSHDQAVR TYQEHKASMH PVTAMLVGKD LKVD | |
Greater than 90.0% as determined by SDS-PAGE. |
LTA4H Protein | |
Research Use Only | |
LTA4H | |
Human | |
His | |
LTA4H protein solution (0.25 mg/ml) in Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |