missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LSM12 Protein

A cDNA sequence encoding the LSM12 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12590-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950919

  • £99.15 / 2µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

124801
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAAPPGE YFSVGSQVSC RTCQEQRLQG EVVAFDYQSK MLALKCPSSS GKPNHADILL INLQYVSEVE IINDRTETPP PLASLNVSKL ASKARTEKEE KLSQAYAISA GVSLEGQQLF QTIHKTIKDC KWQEKNIVVM EEVVITPPYQ VENCKGKEGS ALSHVRKIVE KHFRDVESQK ILQRSQAQQP QKEAALSS
Greater than 90% as determined by SDS-PAGE.
LSM12 Protein
Research Use Only
LSM12
Human
His
LSM12 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers