missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LRP2BP Protein

A cDNA sequence encoding the LRP2BP was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12584-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960899

  • £155.00 / 20µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LRP2BP Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMKLTSEK LPKNPFYASV SQYAAKNQKF FQWKKEKTDY THANLVDKAL QLLKERILKG DTLAYFLRGQ LYFEEGWYEE ALEQFEEIKE KDHQATYQLG VMYYDGLGTT LDAEKGVDYM KKILDSPCPK ARHLKFAAAY NLGRAYYEGK GVKRSNEEAE RLWLIAADNG NPKASVKAQS MLGLYYSTKE PKELEKAFYW HSEACGNGNL ESQGALGLMY LYGQGIRQDT EAALQCLREA AERGNVYAQG NLVEYYYKMK FFTKCVAFSK RIADYDEVHD IPMIAQVTDC LPEFIGRGMA MASFYHARCL QLGLGITRDE TTAKHYYSKA CRLNPALADE LHSLLIRQRI
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The LRP2BP solution (0.5 mg/1ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers