missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRP2BP Protein
A cDNA sequence encoding the LRP2BP was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12584-1mg
Additional Details : Weight : 0.01000kg
Specifications
LRP2BP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKLTSEK LPKNPFYASV SQYAAKNQKF FQWKKEKTDY THANLVDKAL QLLKERILKG DTLAYFLRGQ LYFEEGWYEE ALEQFEEIKE KDHQATYQLG VMYYDGLGTT LDAEKGVDYM KKILDSPCPK ARHLKFAAAY NLGRAYYEGK GVKRSNEEAE RLWLIAADNG NPKASVKAQS MLGLYYSTKE PKELEKAFYW HSEACGNGNL ESQGALGLMY LYGQGIRQDT EAALQCLREA AERGNVYAQG NLVEYYYKMK FFTKCVAFSK RIADYDEVHD IPMIAQVTDC LPEFIGRGMA MASFYHARCL QLGLGITRDE TTAKHYYSKA CRLNPALADE LHSLLIRQRI | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LRP2BP solution (0.5 mg/1ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |