missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LIN7A Protein
A cDNA sequence encoding the LIN7A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12573-1mg
Additional Details : Weight : 0.01000kg
Specifications
LIN7A Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LIN7A protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLKPSVT SAPTADMATL TVVQPLTLDR DVARAIELLE KLQESGEVPV HKLQSLKKVL QSEFCTAIRE VYQYMHETIT VNGCPEFRAR ATAKATVAAF AASEGHSHPR VVELPKTDEG LGFNVMGGKE QNSPIYISRI IPGGVAERHG GLKRGDQLLS VNGVSVEGEH HEKAVELLKA AKDSVKLVVR YTPKVLEEME ARFEKLRTAR RRQQQQLLIQ QQQQQQQQQT QQNHMS |