missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIN28 Protein

A cDNA sequence encoding the LIN28 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12571-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950849

  • £2425.00 / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

LIN28 Protein
Research Use Only
Recombinant Protein
E. coli
GPSVSNQQFA GGCAKAAEEA PEEAPEDAAR AADEPQLLHG AGICKWFNVR MGFGFLSMTA RAGVALDPPV DVFVHQSKLH MEGFRSLKEG EAVEFTFKKS AKGLESIRVT GPGGVFCIGS ERRPKGKSMQ KRRSKGDRCY NCGGLDHHAK ECKLPPQPKK CHFCQSISHM VASCPLKAQQ GPSAQGKPTY FREEEEEIHS PTLLPEAQNG GYGRKKRRQR RR
Greater than 90% as determined by SDS-PAGE (coomassie staining).
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The LIN28 protein solution was formulated in PBS with 50mM arginine
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers