missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LILRB1/CD85j/ILT2 His Avi-tag Protein
Biotinylated
Brand: R&D Systems™ AVI8989-050
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Human LILRB1/CD85j/ILT2 Antibody (Catalog # MAB20172 ) is immobilized at 0.2 μg/mL (100 μL/well), Biotinylated Recombinant Human LILRB1/CD85j/ILT2 His-tag Avi-tag (Catalog # AVI8989) binds with an ED50 of 0.600-9.00 ng/mL.Specifications
Q8NHL6.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
M.W. Observed: 70-80 kDa, under reducing conditions., M.W. Theoretical: 50 kDa | |
Chinese Hamster Ovary cell line, CHO-derived human LILRB1/CD85j/ILT2 protein Human LILRB1/CD85j/ILT2 (Gly24-Arg457) (N-terminus) Accession # Q8NHL6.1 6-His tag Avi-tag (C-terminus) | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Biotin | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining. |
Bioactivity | |
10859 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
CD85 antigen-like family member J, CD85J, CD85j antigen, Ig-like transcript 2, ILT2, ILT-2, ILT2FLJ37515, Immunoglobulin-like transcript 2, leukocyte Ig-like receptor-1, Leukocyte immunoglobulin-like receptor 1, leukocyte immunoglobulin-like receptor subfamily B member 1, leukocyte immunoglobulin-like receptor subfamily B member 1 soluble isoform, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LIR1, LIR1LIR-1MIR-7CD85MIR7CD85 antigen, member 1, MIR7, Monocyte/macrophage immunoglobulin-like receptor 7 | |
Reconstitute at 500 μg/mL in PBS. | |
ILT2/CD85j/LILRB1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction