missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LILRA5 Protein

A DNA sequence encoding the Homo sapiens (Human) LILRA5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP2841-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15955569

  • £2016.44 / 1mg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_067073.1
LILRA5 Protein
1 mg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
353514
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LILRA5
Recombinant Protein
Yeast
GNLSKATLWA EPGSVISRGN SVTIRCQGTL EAQEYRLVKE GSPEPWDTQN PLEPKNKARF SIPSMTEHHA GRYRCYYYSP AGWSEPSDPL ELVVTGFYNK PTLSALPSPV VTSGENVTLQ CGSRLRFDRF ILTEEGDHKL SWTLDSQLTP SGQFQALFPV GPVTPSHRWM LRCYGSRRHI LQVWSEPSDL LEIPVSGAAD NLSPSQNKSD SGTASHLQDY AVENLIR
Videos
SDS
Documents
Certificates
Special Offers

Special Offers