missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGALS13 Protein
A cDNA sequence encoding the LGALS13 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | LGALS13 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980809
|
enQuireBio™
QP12559-20UG |
20 μg |
£99.15
20µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960809
|
enQuireBio™
QP12559-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970809
|
enQuireBio™
QP12559-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
LGALS13 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
LGALS13 protein solution (0.5 mg/ml) is formulated in 1xPBS buffer pH 7.4. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
macglvasnl nlkpgeclrv rgevapdaks fvlnlgkdsn nlclhfnprf nahgdantiv cnskdggawg teqreavfpf qpgsvaevci tfdqanltvk lpdgyefkfp nrlnleainy maadgdfkik cvafd | |
Greater than 90.0% as determined by SDS-PAGE. |