missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leuprorelin Protein
Recombinant Protein
Brand: enQuireBio™ QP12555-100mg
Additional Details : Weight : 0.01000kg
Specifications
Leuprorelin Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
pGlu-His-Trp-Ser-Tyr-D-Leu-Leu-Arg-Pro-NHEt | |
Greater than 98.0% as determined by RP-HPLC. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The Leuprorelin was lyophilized from a concentrated (1 mg/ml) solution with no additives. |