missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Leptin qA Protein
A cDNA sequence encoding the Leptin qA was constructed and used to recombinantly synthesize the protein.
£99.15 - £1855.00
Specifications
Name | Leptin qA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Leptin Quadruple Antagonist Mutant is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transected with the long form of human Leptin receptor. It also inhibits various Leptin effects in several in vitro bioassays. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15955078
|
enQuireBio™
QP10758-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975078
|
enQuireBio™
QP10758-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965078
|
enQuireBio™
QP10758-1MG |
1 mg |
£1855.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Leptin qA Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3. |
Research Use Only | |
Leptin Quadruple Antagonist Mutant is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transected with the long form of human Leptin receptor. It also inhibits various Leptin effects in several in vitro bioassays. | |
Human | |
Untagged | |
Greater than 98.0% as determined by:(a) Gel filtration analysis. (b) Analysis by SDS-PAGE. |