missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LANCL1 Protein
A cDNA sequence encoding the LANCL1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12535-1mg
Weitere Details : Weight : 0.01000kg
Spezifikation
LANCL1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAQRAFP NPYADYNKSL AEGYFDAAGR LTPEFSQRLT NKIRELLQQM ERGLKSADPR DGTGYTGWAG IAVLYLHLYD VFGDPAYLQL AHGYVKQSLN CLTKRSITFL CGDAGPLAVA AVLYHKMNNE KQAEDCITRL IHLNKIDPHA PNEMLYGRIG YIYALLFVNK NFGVEKIPQS HIQQICETIL TSGENLARKR NFTAKSPLMY EWYQEYYVGA AHGLAGIYYY LMQPSLQVSQ GKLHSLVKPS VDYVCQLKFP SGNYPPCIGD NRDLLVHWCH GAPGVIYMLI QAYKVFREEK YLCDAYQCAD VIWQYGLLKK GYGLCHGSAG NAYAFLTLYN LTQDMKYLYR ACKFAEWCLE YGEHGCRTPD TPFSLFEGMA GTIYFLADLL VPTKARFPAF EL | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LANCL1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 0.15M NaCl, 20% glycerol and 1mM DTT. |