missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAMP1 Protein
A cDNA sequence encoding the LAMP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12529-1mg
Additional Details : Weight : 0.01000kg
Specifications
LAMP1 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPAMFMVKN GNGTACIMAN FSAAFSVNYD TKSGPKNMTF DLPSDATVVL NRSSCGKENT SDPSLVIAFG RGHTLTLNFT RNATRYSVQL MSFVYNLSDT HLFPNASSKE IKTVESITDI RADIDKKYRC VSGTQVHMNN VTVTLHDATI QAYLSNSSFS RGETRCEQDR PSPTTAPPAPPSPSPSPVPK SPSVDKYNVS GTNGTCLLAS MGLQLNLTYE RKDNTTVTRL LNINPNKTSA SGSCGAHLVT LELHSEGTTV LLFQFGMNAS SSRFFLQGIQ LNTILPDARD PAFKAANGSL RALQATVGNS YKCNAEEHVR VTKAFSVNIF KVWVQAFKVE GGQFGSVEEC LLDENSMHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
LAMP1 protein solution (1 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |