missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human L2HGDH, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) L2HGDH, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8068-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002481

  • £369.88 / 100µg

Estimated Shipment
02-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_079160.1
L2HGDH, mitochondrial Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
79944
Greater than 90% as determined by SDS-PAGE.
Research Use Only
L2HGDH
Recombinant Protein
E. coli
VIVGGGIVGL ASARALILRH PSLSIGVLEK EKDLAVHQTG HNSGVIHSGI YYKPESLKAK LCVQGAALLY EYCQQKGISY KQCGKLIVAV EQEEIPRLQA LYEKGLQNGV PGLRLIQQED IKKKEPYCRG LMAIDCPHTG IVDYRQVALS FAQDFQEAGG SVLTNFEVKG IEMAKESPSR SIDGMQYPIV IKNTKGEEIR CQYVVTCAGL YSDRISELSG CTPDPRIVPF RGDYLLLKPE KCYLVKGNIY PVPDSRFPFL GVHFTPRMDG SIWLGPNAVL AFKREGYRPF DFSATDVMDI IINSGLIKLA SQNFSYGVTE MYKACFLGAT VKYLQKFIPE ITISDILRGP AGVRAQALDR DGNLVEDFVF DAGVGDIGNR ILHVRNAPSP AATSSIAISG MIADEVQQRF EL
Videos
Documents