missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRG2 Protein
A DNA sequence encoding the Homo sapiens (Human) KLRG2, was expressed in the hosts and tags indicated.
£183.90 - £1963.71
Specifications
Accession Number | NP_940910.1 |
---|---|
Gene ID (Entrez) | 346689 |
Name | KLRG2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007891
|
enQuireBio™
QP9236-YE-100UG |
100 μg |
£467.00
100µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047891
|
enQuireBio™
QP9236-YE-500UG |
500 μg |
£1265.00
500µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017891
|
enQuireBio™
QP9236-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037891
|
enQuireBio™
QP9236-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16057891
|
enQuireBio™
QP9236-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16027891
|
enQuireBio™
QP9236-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_940910.1 | |
KLRG2 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
346689 | |
Research Use Only | |
KLRG2 | |
Recombinant Protein | |
Yeast | |
MEESWEAAPG GQAGAELPME PVGSLVPTLE QPQVPAKVRQ PEGPESSPSP AGAVEKAAGA GLEPSSKKKP PSPRPGSPRV PPLSLGYGVC PEPPSPGPAL VKLPRNGEAP GAEPAPSAWA PMELQVDVRV KPVGAAGGSS TPSPRPSTRF LKVPVPESPA FSRHADPAHQ LLLRAPSQGG TWGRRSPLAA ARTESGCDAE GRASPAEGSA GSPGSPTCCR CKELGLEKED AALLPRAGLD GDEKLPRAVT LTGLPMYVKS LYWALAFMAV LLAVSGVVIV VLASRAGARC QQCPPGWVLS EEHCYYFSAE AQAWEASQAF CSAYHATLPL LSHTQDFLGR YPVSRHSWVG AWRGPQGWHW IDEAPLPPQL LPEDGEDNLD INCGALEEGT LVAANCSTPR PWVCAKGTQ | |
Greater than 90% as determined by SDS-PAGE. |