missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLHDC8B Protein

A cDNA sequence encoding the KLHDC8B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12501-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960619

  • £155.00 / 20µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

KLHDC8B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMSAGGGR AFAWQVFPPM PTCRVYGTVA HQDGHLLVLG GCGRAGLPLD TAETLDMASH TWLALAPLPT ARAGAAAVVL GKQVLVVGGV DEVQSPVAAV EAFLMDEGRW ERRATLPQAA MGVATVERDG MVYALGGMGP DTAPQAQVRV YEPRRDCWLS LPSMPTPCYG ASTFLHGNKI YVLGGRQGKL PVTAFEAFDL EARTWTRHPS LPSRRAFAGC AMAEGSVFSL GGLQQPGPHN FYSRPHFVNT VEMFDLEHGS WTKLPRSLRM RDKRADFVVG SLGGHIVAIG GLGNQPCPLG SVESFSLARR RWEALPAMPT ARCSCSSLQA GPRLFVIGGV AQGPSQAVEA LCLRDGV
Greater than 85.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
KLHDC8B protein solution (0.5 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers