missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kinetochore-associated protein DSN1 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Kinetochore-associated protein DSN1 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8173-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003061

  • £223.39 / 10µg

Estimated Shipment
06-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001138787.1
Kinetochore-associated protein DSN1 homolog Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
79980
Greater than 80% as determined by SDS-PAGE.
Research Use Only
DSN1
Recombinant Protein
E. coli
MTSVTRSEII DEKGPVMSKT HDHQLESSLS PVEVFAKTSA SLEMNQGVSE ERIHLGSSPK KGGNCDLSHQ ERLQSKSLHL SPQEQSASYQ DRRQSWRRAS MKETNRRKSL HPIHQGITEL SRSISVDLAE SKRLGCLLLS SFQFSIQKLE PFLRDTKGFS LESFRAKASS LSEELKHFAD GLETDGTLQK CFEDSNGKAS DFSLEASVAE MKEYITKFSL ERQTWDQLLL HYQQEAKEIL SRGSTEAKIT EVKVEPMTYL GSSQNEVLNT KPDYQKILQN QSKVFDCMEL VMDELQGSVK QLQAFMDEST QCFQKVSVQL GKRSMQQLDP SPARKLLKLQ LQNPPAIHGS GSGSCQ
Videos
Documents