missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kinetochore-associated protein DSN1 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Kinetochore-associated protein DSN1 homolog, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001138787.1 |
---|---|
Gene ID (Entrez) | 79980 |
Name | Kinetochore-associated protein DSN1 homolog Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093051
|
enQuireBio™
QP8173-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013061
|
enQuireBio™
QP8173-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003061
|
enQuireBio™
QP8173-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023061
|
enQuireBio™
QP8173-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033061
|
enQuireBio™
QP8173-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043061
|
enQuireBio™
QP8173-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001138787.1 | |
Kinetochore-associated protein DSN1 homolog Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
79980 | |
Research Use Only | |
DSN1 | |
Recombinant Protein | |
E. coli | |
MTSVTRSEII DEKGPVMSKT HDHQLESSLS PVEVFAKTSA SLEMNQGVSE ERIHLGSSPK KGGNCDLSHQ ERLQSKSLHL SPQEQSASYQ DRRQSWRRAS MKETNRRKSL HPIHQGITEL SRSISVDLAE SKRLGCLLLS SFQFSIQKLE PFLRDTKGFS LESFRAKASS LSEELKHFAD GLETDGTLQK CFEDSNGKAS DFSLEASVAE MKEYITKFSL ERQTWDQLLL HYQQEAKEIL SRGSTEAKIT EVKVEPMTYL GSSQNEVLNT KPDYQKILQN QSKVFDCMEL VMDELQGSVK QLQAFMDEST QCFQKVSVQL GKRSMQQLDP SPARKLLKLQ LQNPPAIHGS GSGSCQ | |
Greater than 80% as determined by SDS-PAGE. |