missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kinetochore-associated protein DSN1 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Kinetochore-associated protein DSN1 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8173-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013061

  • £2480.00 / 1mg
Estimated Shipment: 09-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138787.1
Kinetochore-associated protein DSN1 homolog Protein
Research Use Only
DSN1
Recombinant Protein
E. coli
MTSVTRSEII DEKGPVMSKT HDHQLESSLS PVEVFAKTSA SLEMNQGVSE ERIHLGSSPK KGGNCDLSHQ ERLQSKSLHL SPQEQSASYQ DRRQSWRRAS MKETNRRKSL HPIHQGITEL SRSISVDLAE SKRLGCLLLS SFQFSIQKLE PFLRDTKGFS LESFRAKASS LSEELKHFAD GLETDGTLQK CFEDSNGKAS DFSLEASVAE MKEYITKFSL ERQTWDQLLL HYQQEAKEIL SRGSTEAKIT EVKVEPMTYL GSSQNEVLNT KPDYQKILQN QSKVFDCMEL VMDELQGSVK QLQAFMDEST QCFQKVSVQL GKRSMQQLDP SPARKLLKLQ LQNPPAIHGS GSGSCQ
Greater than 80% as determined by SDS-PAGE.
79980
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers