missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KCTD4 Protein

A cDNA sequence encoding the KCTD4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12481-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950559

  • £99.15 / 2µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

386618
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMERKINR REKEKEYEGK HNSLEDTDQG KNCKSTLMTL NVGGYLYITQ KQTLTKYPDT FLEGIVNGKI LCPFDADGHY FIDRDGLLFR HVLNFLRNGE LLLPEGFREN QLLAQEAEFF QLKGLAEEVK SRWEKEQLTP RETTFLEITD NHDRSQGLRI FCNAPDFISK IKSRIVLVSK SRLDGFPEEF SISSNIIQFK YFIKSENGTR LVLKEDNTFV CTLETLKFEA IMMALKCGFR LLTSLDCSKG SIVHSDALHF IK
Greater than 85% as determined by SDS-PAGE.
KCTD4 Protein
Research Use Only
KCTD4
Human
His
KCTD4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 40% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers