missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCTD4 Protein
A cDNA sequence encoding the KCTD4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12481-10ug
Additional Details : Weight : 0.01000kg
Specifications
386618 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMERKINR REKEKEYEGK HNSLEDTDQG KNCKSTLMTL NVGGYLYITQ KQTLTKYPDT FLEGIVNGKI LCPFDADGHY FIDRDGLLFR HVLNFLRNGE LLLPEGFREN QLLAQEAEFF QLKGLAEEVK SRWEKEQLTP RETTFLEITD NHDRSQGLRI FCNAPDFISK IKSRIVLVSK SRLDGFPEEF SISSNIIQFK YFIKSENGTR LVLKEDNTFV CTLETLKFEA IMMALKCGFR LLTSLDCSKG SIVHSDALHF IK | |
Greater than 85% as determined by SDS-PAGE. |
KCTD4 Protein | |
Research Use Only | |
KCTD4 | |
Human | |
His | |
KCTD4 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 40% glycerol and 1mM DTT. |