missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human JNK2 / MAPK9 Protein
A DNA sequence encoding the Homo sapiens (Human) JNK2 / MAPK9, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6342-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001128516.1 | |
JNK2 / MAPK9 Protein | |
Research Use Only | |
MAPK9 | |
Recombinant Protein | |
E. coli | |
MSDSKCDSQF YSVQVADSTF TVLKRYQQLK PIGSGAQGIV CAAFDTVLGI NVAVKKLSRP FQNQTHAKRA YRELVLLKCV NHKNIISLLN VFTPQKTLEE FQDVYLVMEL MDANLCQVIH MELDHERMSY LLYQMLCGIK HLHSAGIIHR DLKPSNIVVK SDCTLKILDF GLARTACTNF MMTPYVVTRY YRAPEVILGM GYKENVDIWS VGCIMGELVK GCVIFQGTDH IDQWNKVIEQ LGTPSAEFMK KLQPTVRNYV ENRPKYPGIK FEELFPDWIF PSESERDKIK TSQARDLLSK MLVIDPDKRI SVDEALRHPY ITVWYDPAEA EAPPPQIYDA QLEEREHAIE EWKELIYKEV MDWEERSKNG VVKDQPSDAA VSSNATPSQS SSINDISSMS TEQTLASDTD SSLDASTGPL EGCR | |
Greater than 90% as determined by SDS-PAGE. |
5601 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |