missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human JAM3 Protein

A cDNA sequence encoding the JAM3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12468-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950519

  • £155.00 / 10µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

83700
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMVNLKSS NRTPVVQEFE SVELSCIITD SQTSDPRIEW KKIQDEQTTY VFFDNKIQGD LAGRAEILGK TSLKIWNVTR RDSALYRCEV VARNDRKEID EIVIELTVQV KPVTPVCRVP KAVPVGKMAT LHCQESEGHP RPHYSWYRND VPLPTDSRAN PRFRNSSFHL NSETGTLVFT AVHKDDSGQY YCIASNDAGS ARCEEQEMEV YDLN
Greater than 90% as determined by SDS-PAGE.
JAM3 Protein
Research Use Only
JAM3
Human
His
The JAM3 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 2mM EDTA, 5mM DTT and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers