missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interleukin enhancer-binding factor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Interleukin enhancer-binding factor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8309-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003931

  • £145.46 / 10µg

Estimated Shipment
07-10-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_001254738.1
Interleukin enhancer-binding factor 2 Protein
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
3608
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ILF2
Recombinant Protein
E. coli
MRGDRGRGRG GRFGSRGGPG GGFRPFVPHI PFDFYLCEMA FPRVKPAPDE TSFSEALLKR NQDLAPNSAE QASILSLVTK INNVIDNLIV APGTFEVQIE EVRQVGSYKK GTMTTGHNVA DLVVILKILP TLEAVAALGN KVVESLRAQD PSEVLTMLTN ETGFEISSSD ATVKILITTV PPNLRKLDPE LHLDIKVLQS ALAAIRHARW FEENASQSTV KVLIRLLKDL RIRFPGFEPL TPWILDLLGH YAVMNNPTRQ PLALNVAYRR CLQILAAGLF LPGSVGITDP CESGNFRVHT VMTLEQQDMV CYTAQTLVRI LSHGGFRKIL GQEGDASYLA SEISTWDGVI VTPSEKAYEK PPEKKEGEEE EENTEEPPQG EEEESMETQE
Videos
Documents