missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha 2b Protein
Recombinant Protein
Brand: enQuireBio™ QP5213-500ug
Additional Details : Weight : 0.01000kg
Specifications
Interferon alpha 2b Protein | |
Research Use Only | |
IFNA2 | |
Recombinant Protein | |
E. coli | |
MCDLPQTHSL GSRRTLMLLA QMRRISLFSC LKDRHDFGFP QEEFGNQFQK AETIPVLHEM IQQIFNLFST KDSSAAWDET LLDKFYTELY QQLNDLEACV IQGVGVTETP LMKEDSILAV RKYFQRITLY LKEKKYSPCA WEVVRAEIMR SFSLSTNLQE SLRSKE | |
Greater than 95% as determined by SDS-PAGE. |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity in the Viral CPE assay using EMC virus on A549 cells assay. The bioactivity acceptance criteria is set at 2.0 x 10^8 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |