missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Insulin Protein
A cDNA sequence encoding the Insulin was constructed and used to recombinantly synthesize the protein.
£99.15 - £1100.00
Specifications
Gene ID (Entrez) | 3630 |
---|---|
Name | Insulin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | INS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15905008
|
enQuireBio™
QP10732-2MG |
2 mg |
£99.15
2mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964998
|
enQuireBio™
QP10732-10MG |
10 mg |
£155.00
10mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994998
|
enQuireBio™
QP10732-25MG |
25 mg |
£136.00
25mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954998
|
enQuireBio™
QP10732-100MG |
100 mg |
£660.00
100mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984998
|
enQuireBio™
QP10732-250MG |
250 mg |
£357.00
250mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974998
|
enQuireBio™
QP10732-1GR |
1 g |
£1100.00
1g |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
3630 | |
Research Use Only | |
INS | |
Recombinant Protein | |
Untagged | |
The protein was lyophilized from 50mM Sodium Chloride solution. |
Insulin Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
27.5 units/mg. Insulin has been evaluated in cell culture (Human Foreskin Fibroblasts). The effective concentration range for use in defined media is 1-5ug/ml. However, the optimum concentration range for a particular application must be determined by the investigator. | |
Human | |
MALWMRLLPL LALLALWGPD PAAAFVNQHL CGSHLVEALY LVCGERGFFY TPKTRREAED LQVGQVELGG GPGAGSLQPL ALEGSLQKRG IVEQCCTSIC SLYQLENYCN | |
Greater than 95.0% as determined by SDS-PAGE. |