missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human INHBC Protein
A cDNA sequence encoding the INHBC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12447-10ug
Additional Details : Weight : 0.01000kg
Specifications
3626 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGIDCQGG SRMCCRQEFF VDFREIGWHD WIIQPEGYAM NFCIGQCPLH IAGMPGIAAS FHTAVLNLLK ANTAAGTTGG GSCCVPTARR PLSLLYYDRD SNIVKTDIPD MVVEACGCS | |
Greater than 85% as determined by SDS-PAGE. |
INHBC Protein | |
Research Use Only | |
INHBC | |
Human | |
His | |
The INHBC solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |