missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Inactive tyrosine-protein kinase 7 Protein

A DNA sequence encoding the Homo sapiens (Human) Inactive tyrosine-protein kinase 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9763-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048681

  • £405.00 / 100µg

Estimated Shipment: 25-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001257327.1
Inactive tyrosine-protein kinase 7 Protein
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
5754
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PTK7
Recombinant Protein
Yeast
AIVFIKQPSS QDALQGRRAL LRCEVEAPGP VHVYWLLDGA PVQDTERRFA QGSSLSFAAV DRLQDSGTFQ CVARDDVTGE EARSANASFN IKWIEAGPVV LKHPASEAEI QPQTQVTLRC HIDGHPRPTY QWFRDGTPLS DGQSNHTVSS KERNLTLRPA GPEHSGLYSC CAHSAFGQAC SSQNFTLSIA DESFARVVLA PQDVVVARYE EAMFHCQFSA QPPPSLQWLF EDETPITNRS RPPHLRRATV FANGSLLLTQ VRPRNAGIYR CIGQGQRGPP IILEATLHLA EIEDMPLFEP RVFTAGSEER VTCLPPKGLP EPSVWWEHAG VRLPTHGRVY QKGHELVLAN IAESDAGVYT CHAANLAGQR RQDVNITVAT VPSWLKKPQD SQLEEGKPGY LDCLTQATPK PTVVWYRNQM LISEDSRFEV FKNGTLRINS VEVYDGTWYR CMSSTPAGSI EAQARVQVLE KLKFTPPPQP QQCMEFDKEA TVPCSATGRE KPTIKWERAD GSSLPEWVTD NAGTLHFARV TRDDAGNYTC IASNGPQGQI RAHVQLTVAV FITFKVEPER TTVYQGHTAL LQCEAQGDPK PLIQWKGKDR ILDPTKLGPR MHIFQNGSLV IHDVAPEDSG RYTCIAGNSC NIKHTEAPLY VVDKPVPEES EGPGSPPPYK MIQT
Videos
SDS
Documents
Certificates