missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IMPACT Protein
A cDNA sequence encoding the IMPACT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12434-1mg
Additional Details : Weight : 0.01000kg
Specifications
IMPACT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEGDAG SDQRQNEEIE AMAAIYGEEW CVIDDCAKIF CIRISDDIDD PKWTLCLQVM LPNEYPGTAP PIYQLNAPWL KGQERADLSN SLEEIYIQNI GESILYLWVE KIRDVLIQKS QMTEPGPDVK KKTEEEDVEC EDDLILACQP ESSVKALDFD ISETRTEVEV EELPPIDHGI PITDRRSTFQ AHLAPVVCPK QVKMVLSKLY ENKKIASATH NIYAYRIYCE DKQTFLQDCE DDGETAAGGR LLHLMEILNV KNVMVVVSRW YGGILLGPDR FKHINNCARN ILVEKNYTNS PEESSKALGK NKKVRKDKKR NEH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
IMPACT protein solution (1 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |