missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL6 / Interleukin-6 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5235-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965749

  • £3092.23 / 1mg
Estimated Shipment: 13-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000591.1
IL6 / Interleukin-6 Protein
1 mg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 250 pg/mL in the 7TD1 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
3569
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL6
Recombinant Protein
E. coli
MPVPPGEDSK DVAAPHRQPL TSSERIDKQI RYILDGISAL RKETCNKSNM CESSKEALAE NNLNLPKMAE KDGCFQSGFN EETCLVKIIT GLLEFEVYLE YLQNRFESSE EQARAVQMST KVLIQFLQKK AKNLDAITTP DPTTNASLLT KLQAQNQWLQ DMTTHLILRS FKEFLQSSLR ALRQM
Videos
SDS
Documents
Certificates
Special Offers

Special Offers