missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL4R Protein

A cDNA sequence encoding the IL4R was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12425-0.1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960379

  • £990.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

3566
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
MKVLQEPTCV SDYMSISTCE WKMNGPTNCS TELRLLYQLV FLLSEAHTCI PENNGGAGCV CHLLMDDVVS ADNYTLDLWA GQQLLWKGSF KPSEHVKPRA PGNLTVHTNV SDTLLLTWSN PYPPDNYLYN HLTYAVNIWS ENDPADFRIY NVTYLEPSLR IAASTLKSGI SYRARVRAWAQCYNTTWSEW SPSTKWHNSY REPFEQHLEH HHHHH
Greater than 95.0% as determined by SDS-PAGE.
IL4R Protein
Research Use Only
IL4R
Human
His
IL4R protein solution (0.25 mg/ml) contains Phosphate buffered saline (pH 7.4).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers