missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL4 / Interleukin-4 Protein

A DNA sequence encoding the Homo sapiens (Human) IL4 / Interleukin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8557-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16085261

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000580.1
IL4 / Interleukin-4 Protein
Research Use Only
IL4
Recombinant Protein
E. coli
HKCDITLQEI IKTLNSLTEQ KTLCTELTVT DIFAASKNTT EKETFCRAAT VLRQFYSHHE KDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE NFLERLKTIM REKYSKCSS
Greater than 90% as determined by SDS-PAGE.
3565
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers