missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL34 Protein

A cDNA sequence encoding the IL34 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10728-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954988

  • £99.15 / 2µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

146433
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
The specific activity was measured by its ability to induce secretion of MCP-1 from PBMCs.
Human
His
The IL34 was lyophilized from a solution containing 20mM Sodium Citrate, pH 3.0 and 75mM NaCl.
IL34 Protein
Research Use Only
IL34
Recombinant Protein
Human (HEK293)
NEPLEMWPLT QNEECTVTGF LRDKLQYRSR LQYMKHYFPI NYKISVPYEG VFRIANVTRL QRAQVSEREL RYLWVLVSLS ATESVQDVLL EGHPSWKYLQ EVETLLLNVQ QGLTDVEVSP QGLTDVEVSP APGPNLKLVR PKALLDNCFR VMELLYCSCC KQSSVLNWQD CEVPSPQSCS PEPSLQYAAT QLYPPPPWSP SSPPHSTGSV RPVRAQGEGL LPHHHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers