missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL21R Protein

A cDNA sequence encoding the IL21R was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12415-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15960349

  • £155.00 / 10µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

50615
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
CPDLVCYTDY LQTVICILEM WNLHPSTLTL TWQDQYEELK DEATSCSLHR SAHNATHATY TCHMDVFHFM ADDIFSVNIT DQSGNYSQEC GSFLLAESIK PAPPFNVTVT FSGQYNISWR SDYEDPAFYM LKGKLQYELQ YRNRGDPWAV SPRRKLISVD SRSVSLLPLE FRKDSSYELQ VRAGPMPGSS YQGTWSEWSD PVIFQTQSEE LKEVEHHHHH H
Greater than 90.0% as determined by SDS-PAGE.
IL21R Protein
Research Use Only
IL21R
Human
His
IL21R protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers