missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL21R Protein
A cDNA sequence encoding the IL21R was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 50615 |
---|---|
Name | IL21R Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IL21R |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15980349
|
enQuireBio™
QP12415-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960349
|
enQuireBio™
QP12415-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15970349
|
enQuireBio™
QP12415-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
50615 | |
Research Use Only | |
IL21R | |
Human | |
His | |
IL21R protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |
IL21R Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
CPDLVCYTDY LQTVICILEM WNLHPSTLTL TWQDQYEELK DEATSCSLHR SAHNATHATY TCHMDVFHFM ADDIFSVNIT DQSGNYSQEC GSFLLAESIK PAPPFNVTVT FSGQYNISWR SDYEDPAFYM LKGKLQYELQ YRNRGDPWAV SPRRKLISVD SRSVSLLPLE FRKDSSYELQ VRAGPMPGSS YQGTWSEWSD PVIFQTQSEE LKEVEHHHHH H | |
Greater than 90.0% as determined by SDS-PAGE. |