missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5245-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965769

  • £470.32 / 500µg
Estimated Shipment: 12-01-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000577.2
IL2 / Interleukin-2 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity in the CTLL-2 cell proliferation assay.
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
3558
Greater than 95% as determined by SDS-PAGE.
Research Use Only
IL2
Recombinant Protein
E. coli
MAPTSSSTKK TQLQLEHLLL DLQMILNGIN NYKNPKLTRM LTFKFYMPKK ATELKHLQCL EEELKPLEEV LNLAQSKNFH LRPRDLISNI NVIVLELKGS ETTFMCEYAD ETATIVEFLN RWITFCQSII STLT
Videos
SDS
Documents
Certificates
Special Offers

Special Offers