missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL13 / ALRH Protein
Recombinant Protein
Brand: enQuireBio™ QP5388-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_002179.2 | |
IL13 / ALRH Protein | |
Research Use Only | |
IL13 | |
Recombinant Protein | |
E. coli | |
MSPGPVPPST ALRELIEELV NITQNQKAPL CNGSMVWSIN LTAGMYCAAL ESLINVSGCS AIEKTQRMLS GFCPHKVSAG QFSSLHVRDT KIEVAQFVKD LLLHLKKLFR EGRQFN | |
Greater than 95% as determined by SDS-PAGE. |
3596 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Human | |
Untagged | |
10 mM sodium citrate, pH 3.0 |