missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-35 Heterodimer Protein
Recombinant Protein
Brand: enQuireBio™ QP5203-100ug
Additional Details : Weight : 0.01000kg
Specifications
IL-35 Heterodimer Protein | |
Research Use Only | |
IL35 | |
Human | |
Untagged | |
10 mM sodium phosphate, 30 mM sodium chloride, pH7.5 |
100 μg | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
EBI3:RKGPP AALTLPRVQC RASRYPIAVD CSWTLPPAPN STSPVSFIAT YRLGMAARGH SWPCLQQTPT STSCTITDVQ LFSMAPYVLN VTAVHPWGSS SSFVPFITEH IIKPDPPEGV RLSPLAERQL QVQWEPPGSW PFPEIFSLKY WIRYKRQGAA RFHRVGPIEA TSFILRAVRP RARYYVQVAA QDLTDYGELS DWSLPATATM SLGK p35:RN LPVATPDPGM FPCLHHSQNL LRAVSNMLQK ARQTLEFYPC TSEEIDHEDI TKDKTSTVEA CLPLELTKNE SCLNSRETSF ITNGSCLASR KTSFMMALCL SSIYEDLKMY QVEFKTMNAK LLMDPKRQIF LDQNMLAVID ELMQALNFNS ETVPQKSSLE EPDFYKTKIK LCILLHAFRI RAVTIDRVMS YLNAS | |
Greater than 95% as determined by SDS-PAGE. |