missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28B Protein
A cDNA sequence encoding the IL 28B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10709-10ug
Additional Details : Weight : 0.01000kg
Specifications
282617 | |
Greater than 95% as obsereved by SDS-PAGE. | |
Research Use Only | |
IFNL3 | |
Recombinant Protein | |
HEK 293 | |
MTGDCMPVLV LMAAVLTVTG AVPVARLRGA LPDARGCHIA QFKSLSPQEL QAFKRAKDAL EESLLLKDCK CRSRLFPRTW DLRQLQVRER PVALEAELAL TLKVLEATAD TDPALGDVLD QPLHTLHHIL SQLRACIQPQ PTAGPRTRGR LHHWLHRLQE APKKESPGCL EASVTFNLFR LLTRDLNCVA SGDLCV |
IL 28B Protein | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus and is typically 0.1-1ng/ml. | |
Human | |
Untagged | |
The IL28B was lyophilized from 1 mg/ml in 1xPBS. |