missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28A Protein
A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.
£99.15 - £155.00
Specifications
Gene ID (Entrez) | 282616 |
---|---|
Name | IL 28A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | IFNL2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970289
|
enQuireBio™
QP12397-HIS-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960289
|
enQuireBio™
QP12397-HIS-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
282616 | |
Research Use Only | |
IFNL2 | |
Human | |
His | |
Lyophilized from a 0.2 ?m filtered solution containing no additives. |
IL 28A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
VPVAR LHGALPDARG CHIAQFKSLS PQELQAFKRAKDALEESLLL KDCRCHSRLF PRTWDLRQLQ VRERPMALEA ELALTLKVLE ATADTDPALV DVLDQPLHTL HHILSQFRAC IQPQPTAGPR TRGRLHHWLY RLQEAPKKES PGCLEASVTFNLFRLLTRDL NCVASGDLCV | |
Greater than 85% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |