missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28A Protein
A cDNA sequence encoding the IL 28A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12397-HIS-10ug
Additional Details : Weight : 0.01000kg
Specifications
282616 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
VPVAR LHGALPDARG CHIAQFKSLS PQELQAFKRAKDALEESLLL KDCRCHSRLF PRTWDLRQLQ VRERPMALEA ELALTLKVLE ATADTDPALV DVLDQPLHTL HHILSQFRAC IQPQPTAGPR TRGRLHHWLY RLQEAPKKES PGCLEASVTFNLFRLLTRDL NCVASGDLCV | |
Greater than 85% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
IL 28A Protein | |
Research Use Only | |
IFNL2 | |
Human | |
His | |
Lyophilized from a 0.2 ?m filtered solution containing no additives. |