missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-27 Ra/WSX-1/TCCR Fc Avi-tag Protein
Biotinylated
Brand: R&D Systems™ AVI1479-050
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human IL-27 (Catalog # 2526-IL/CF ) is immobilized at 2 μg/mL (100 μL/well), Biotinylated Recombinant Human IL-27 R alpha/WSX-1/TCCR Fc Chimera Avi-tag (Catalog # AVI1479) binds with an ED50 of 5.00-60.0 ng/mL.Specifications
Q6UWB1.2 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
M.W. Observed: 99-112 kDa, under reducing conditions., M.W. Theoretical: 81 kDa | |
Human embryonic kidney cell, HEK293-derived human IL-27 R alpha/WSX-1/TCCR protein Human IL-27 R alpha/WSX-1/TCCR (Gln33-Lys516) (N-terminus) Accession # Q6UWB1.2 IEGRMD Human IgG1 (Pro100-Lys330) Avi-tag (C-terminus) | |
<0.10 EU per 1 μg of the protein by the LAL method. | |
Biotin | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining. |
Bioactivity | |
9466 | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. | |
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1 | |
Reconstitute at 500 μg/mL in PBS. | |
IL-27R alpha/WSX-1/TCCR |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction